Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172031.1 | NP_742028.1 | 5 - 6 | 674 | 98 | |
GenBank mRNA | AF393750.1 | - | 5 - 6 | 674 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |