Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100778.1 | NP_001094248.1 | 1 - 2 | 409 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Inventoried Probe spans exons |