Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107791.1 | NP_001101261.1 | 2 - 3 | 165 | 92 | |
XM_006235317.1 | XP_006235379.1 | 11 - 12 | 1125 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |