Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037978.1 | NP_001033067.1 | 5 - 6 | 714 | 146 | |
XM_006255083.1 | XP_006255145.1 | 5 - 6 | 832 | 146 | ||
GenBank mRNA | BC099768.1 | - | 5 - 6 | 714 | 7932140 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |