Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772340.1 | XP_008770562.1 | 6 - 7 | 855 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |