Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255126.2 | XP_006255188.1 | 5 - 6 | 2646 | 84 | |
XM_006255127.2 | XP_006255189.1 | 4 - 5 | 2499 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |