Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271091.1 | NP_001258020.1 | 4 - 5 | 392 | 91 | |
NM_001271092.1 | NP_001258021.1 | 4 - 5 | 392 | 91 | ||
NM_001271093.1 | NP_001258022.1 | 4 - 5 | 339 | 91 | ||
NM_001271094.1 | NP_001258023.1 | 4 - 5 | 339 | 91 | ||
NM_001271095.1 | NP_001258024.1 | 2 - 3 | 257 | 91 | ||
NM_031967.2 | NP_114173.2 | 2 - 3 | 257 | 91 | ||
XM_006255123.2 | XP_006255185.1 | 4 - 5 | 334 | 91 | ||
GenBank mRNA | AF045564.1 | - | 2 - 3 | 189 | 91 | |
AY217029.1 | - | 2 - 3 | 228 | 91 | ||
AY217030.1 | - | 4 - 5 | 339 | 91 | ||
AY217031.1 | - | 4 - 5 | 339 | 91 | ||
AY217032.1 | - | 4 - 5 | 387 | 91 | ||
AY217033.1 | - | 4 - 5 | 387 | 91 | ||
AY255791.1 | - | 2 - 3 | 228 | 91 | ||
FM054107.1 | - | 2 - 3 | 210 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |