Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770584.1 | XP_008768806.1 | 2 - 3 | 537 | 150 | |
XM_008770585.1 | XP_008768807.1 | 3 - 4 | 396 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |