Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dupd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108368.1 | NP_001101838.1 | 2 - 3 | 298 | 94 | |
XM_006251647.2 | XP_006251709.1 | 2 - 3 | 242 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |