Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257349.1 | NP_001244278.1 | 120 | 55 | ||
NM_031754.1 | NP_113942.1 | 137 | 55 | |||
GenBank mRNA | CB609817.1 | - | 137 | 55 | ||
FM112505.1 | - | 120 | 55 | |||
FQ214279.1 | - | 55 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |