Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109518.1 | NP_001102988.1 | 2 - 3 | 361 | 62 | |
GenBank mRNA | FQ210083.1 | - | 2 - 3 | 316 | 62 | |
FQ228795.1 | - | 2 - 3 | 339 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |