Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 25 - 26 | 3424 | 108 | |
GenBank mRNA | AF083341.1 | - | 23 - 24 | 2677 | 108 | |
AF135265.1 | - | 23 - 24 | 3095 | 108 | ||
AY330290.1 | - | 22 - 23 | 2554 | 108 | ||
AY330291.1 | - | 22 - 23 | 2560 | 108 | ||
AY330292.1 | - | 22 - 23 | 2563 | 108 | ||
AY330293.2 | - | 22 - 23 | 2554 | 108 | ||
AY330294.1 | - | 22 - 23 | 2554 | 108 | ||
AY344964.1 | - | 20 - 21 | 2390 | 108 | ||
AY344965.2 | - | 21 - 22 | 2966 | 108 | ||
U40603.1 | - | 25 - 26 | 3424 | 108 | ||
U55995.1 | - | 22 - 23 | 2707 | 108 | ||
U93052.1 | - | 22 - 23 | 2655 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |