Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U55995.1 | - | 27 - 28 | 3490 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |