Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246319.1 | NP_001233248.1 | 1 - 2 | 230 | 111 | |
GenBank mRNA | BC129114.1 | - | 1 - 2 | 230 | 7112235 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |