Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100825.1 | NP_001094295.1 | 4 - 5 | 521 | 96 | |
GenBank mRNA | CB741212.1 | - | 4 - 5 | 378 | 96 | |
FQ215151.1 | - | 4 - 5 | 521 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |