Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107333.1 | NP_001100803.1 | 6 - 7 | 703 | 70 | |
XM_006227781.2 | XP_006227843.1 | 6 - 7 | 4453 | 70 | ||
XM_006227782.2 | XP_006227844.1 | 6 - 7 | 1090 | 70 | ||
XM_008758690.1 | XP_008756912.1 | 6 - 7 | 820 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |