Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082574.1 | NP_001076043.1 | 4 - 5 | 1737 | 175 | |
XM_006227809.2 | XP_006227871.1 | 4 - 5 | 2509 | 175 | ||
XM_006227811.2 | XP_006227873.1 | 4 - 5 | 1950 | 175 | ||
XM_008758678.1 | XP_008756900.1 | 3 - 4 | 2906 | 175 | ||
XM_008758679.1 | XP_008756901.1 | 4 - 5 | 2813 | 175 | ||
GenBank mRNA | BC079475.1 | - | 3 - 4 | 1495 | 175 | |
FQ214620.1 | - | 3 - 4 | 290 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |