Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082574.1 | NP_001076043.1 | 6 - 7 | 1921 | 60 | |
XM_006227809.2 | XP_006227871.1 | 6 - 7 | 2693 | 60 | ||
XM_006227811.2 | XP_006227873.1 | 6 - 7 | 2134 | 60 | ||
XM_008758678.1 | XP_008756900.1 | 5 - 6 | 3090 | 60 | ||
XM_008758679.1 | XP_008756901.1 | 6 - 7 | 2997 | 60 | ||
GenBank mRNA | BC079475.1 | - | 5 - 6 | 1679 | 60 | |
BC161806.1 | - | 4 - 5 | 1717 | 7111194 | 60 | |
FQ214620.1 | - | 5 - 6 | 474 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |