Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082574.1 | NP_001076043.1 | 2 - 3 | 1473 | 95 | |
XM_006227809.2 | XP_006227871.1 | 2 - 3 | 2245 | 95 | ||
XM_006227811.2 | XP_006227873.1 | 2 - 3 | 1686 | 95 | ||
XM_008758678.1 | XP_008756900.1 | 1 - 2 | 2642 | 95 | ||
XM_008758679.1 | XP_008756901.1 | 2 - 3 | 2549 | 95 | ||
GenBank mRNA | BC079475.1 | - | 1 - 2 | 1231 | 95 | |
BC161806.1 | - | 1 - 2 | 1444 | 7111194 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |