Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270854.1 | NP_001257783.1 | 5 - 6 | 534 | 71 | |
NM_012672.3 | NP_036804.2 | 5 - 6 | 534 | 71 | ||
XM_008770500.1 | XP_008768722.1 | 3 - 4 | 446 | 71 | ||
GenBank mRNA | J03819.1 | - | 5 - 6 | 530 | 71 | |
J03933.1 | - | 5 - 6 | 531 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Probe spans exons |