Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033690.1 | NP_001028862.1 | 3 - 4 | 636 | 71 | |
XM_006255357.1 | XP_006255419.1 | 2 - 3 | 226 | 71 | ||
GenBank mRNA | BC101871.1 | - | 3 - 4 | 636 | 5621693 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |