Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108440.1 | NP_001101910.1 | 12 - 13 | 1242 | 74 | |
XM_006255267.2 | XP_006255329.1 | 13 - 14 | 1777 | 74 | ||
XM_008772409.1 | XP_008770631.1 | 10 - 11 | 1383 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons |