Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108440.1 | NP_001101910.1 | 6 - 7 | 614 | 114 | |
XM_006255267.2 | XP_006255329.1 | 7 - 8 | 1149 | 114 | ||
XM_006255268.2 | XP_006255330.1 | 7 - 8 | 1149 | 114 | ||
XM_008772409.1 | XP_008770631.1 | 4 - 5 | 755 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |