Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271295.1 | NP_001258224.1 | 10 - 11 | 3110 | 79 | |
NM_001271296.1 | NP_001258225.1 | 10 - 11 | 3110 | 79 | ||
GenBank mRNA | BC099163.1 | - | 1 - 2 | 91 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |