Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139230.1 | NP_631976.1 | 5 - 6 | 627 | 122 | |
NM_139231.1 | NP_631977.1 | 5 - 6 | 477 | 122 | ||
XM_006233479.2 | XP_006233541.1 | 7 - 8 | 795 | 122 | ||
XM_006233480.2 | XP_006233542.1 | 7 - 8 | 1044 | 122 | ||
XM_006233483.2 | XP_006233545.1 | 6 - 7 | 753 | 122 | ||
XM_006233484.2 | XP_006233546.1 | 6 - 7 | 602 | 122 | ||
XM_006233485.2 | XP_006233547.1 | 5 - 6 | 560 | 122 | ||
XM_008761548.1 | XP_008759770.1 | 6 - 7 | 744 | 122 | ||
GenBank mRNA | AF056034.1 | - | 5 - 6 | 627 | 122 | |
AF056035.1 | - | 5 - 6 | 477 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |