Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759293.1 | XP_008757515.1 | 86 - 87 | 12422 | 109 | |
GenBank mRNA | AF130879.1 | - | 3 - 4 | 215 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |