Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012592.2 | NP_036724.1 | 2 - 3 | 244 | 114 | |
GenBank mRNA | BC088401.1 | - | 2 - 3 | 244 | 7193077 | 114 |
FQ231497.1 | - | 1 - 2 | 86 | 114 | ||
J05031.1 | - | 2 - 3 | 254 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |