Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012598.2 | NP_036730.1 | 3 - 4 | 625 | 71 | |
GenBank mRNA | BC081836.1 | - | 3 - 4 | 625 | 7106121 | 71 |
L03294.1 | - | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |