Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019264.2 | NP_062137.1 | 3 - 4 | 449 | 74 | |
XM_006243712.2 | XP_006243774.1 | 4 - 5 | 634 | 74 | ||
XM_006243713.2 | XP_006243775.1 | 4 - 5 | 448 | 74 | ||
XM_008766478.1 | XP_008764700.1 | 3 - 4 | 399 | 74 | ||
GenBank mRNA | AF533978.1 | - | 3 - 4 | 453 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |