Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113740.1 | NP_001107212.1 | 6 - 7 | 633 | 132 | |
NM_022616.2 | NP_072138.2 | 5 - 6 | 630 | 132 | ||
GenBank mRNA | AF142759.1 | - | 5 - 6 | 597 | 132 | |
BC128738.1 | - | 6 - 7 | 531 | 7381331 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |