Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024000.1 | NP_076490.1 | 6 - 7 | 696 | 64 | |
GenBank mRNA | L22557.1 | - | 6 - 7 | 696 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |