Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031542.2 | NP_113730.2 | 21 - 22 | 8719 | 88 | |
GenBank mRNA | AF322646.1 | - | 1 - 2 | 89 | 88 | |
U89653.1 | - | 21 - 22 | 8718 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |