Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031569.3 | NP_113757.1 | 1 - 2 | 207 | 57 | |
XM_006235736.2 | XP_006235798.1 | 2 - 3 | 297 | 57 | ||
XM_006235739.2 | XP_006235801.1 | 2 - 3 | 297 | 57 | ||
GenBank mRNA | L28144.1 | - | 1 - 2 | 138 | 57 | |
L33916.1 | - | 1 - 2 | 72 | 57 | ||
U01913.1 | - | 1 - 2 | 147 | 57 | ||
U05239.1 | - | 1 - 2 | 149 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |