Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 9 - 10 | 1213 | 97 | |
GenBank mRNA | BC062019.1 | - | 9 - 10 | 1123 | 5620971 | 97 |
D83522.1 | - | 9 - 10 | 1087 | 97 | ||
FQ225947.1 | - | 8 - 9 | 872 | 97 | ||
FQ226273.1 | - | 8 - 9 | 872 | 97 | ||
FQ232987.1 | - | 8 - 9 | 873 | 97 | ||
FQ233836.1 | - | 8 - 9 | 872 | 97 | ||
U77918.1 | - | 9 - 10 | 1213 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |