Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052979.1 | NP_443211.1 | 2 - 3 | 252 | 116 | |
GenBank mRNA | AB045587.1 | - | 2 - 3 | 252 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |