Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133548.1 | NP_598232.1 | 1 - 2 | 78 | 82 | |
XM_006244289.2 | XP_006244351.1 | 1 - 2 | 197 | 82 | ||
XM_006244290.2 | XP_006244352.1 | 1 - 2 | 198 | 82 | ||
XM_006244291.2 | XP_006244353.1 | 1 - 2 | 197 | 82 | ||
GenBank mRNA | AF355102.1 | - | 1 - 2 | 78 | 82 | |
BC071178.2 | - | 1 - 2 | 141 | 7100147 | 82 | |
BC087695.1 | - | 1 - 2 | 126 | 7312553 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |