Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 3 - 4 | 221 | 117 | |
XM_006244296.2 | XP_006244358.1 | 3 - 4 | 117 | |||
XM_008766746.1 | XP_008764968.1 | 4 - 5 | 117 | |||
XM_008766747.1 | XP_008764969.1 | 117 | ||||
XM_008766748.1 | XP_008764970.1 | 4 - 5 | 117 | |||
XM_008766749.1 | XP_008764971.1 | 3 - 4 | 117 | |||
XM_008766750.1 | XP_008764972.1 | 3 - 4 | 117 | |||
XM_008766751.1 | XP_008764973.1 | 4 - 5 | 117 | |||
XM_008766752.1 | XP_008764974.1 | 3 - 4 | 117 | |||
XM_008766753.1 | XP_008764975.1 | 3 - 4 | 117 | |||
GenBank mRNA | AF482705.1 | - | 3 - 4 | 221 | 117 | |
BC086988.1 | - | 3 - 4 | 7192831 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |