Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145723.1 | NP_663775.1 | 2 - 3 | 251 | 70 | |
XM_006238024.2 | XP_006238086.1 | 3 - 4 | 329 | 70 | ||
GenBank mRNA | AF115435.1 | - | 2 - 3 | 251 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |