Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153317.1 | NP_695229.1 | 1 - 2 | 189 | 70 | |
GenBank mRNA | BC158579.1 | - | 1 - 2 | 215 | 9091689 | 70 |
U53475.1 | - | 1 - 2 | 189 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |