Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178105.2 | NP_835206.1 | 1 - 2 | 62 | 98 | |
GenBank mRNA | AB089242.1 | - | 1 - 2 | 41 | 98 | |
BC088862.1 | - | 1 - 2 | 62 | 7313006 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |