Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198764.1 | NP_942059.1 | 6 - 7 | 1367 | 95 | |
XM_008771225.1 | XP_008769447.1 | 5 - 6 | 1484 | 95 | ||
GenBank mRNA | AF515735.1 | - | 6 - 7 | 1367 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |