Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY093422.1 | - | 1 - 2 | 84 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |