Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199230.1 | NP_954700.1 | 4 - 5 | 828 | 78 | |
XM_006242315.2 | XP_006242377.1 | 4 - 5 | 1672 | 78 | ||
XM_008765678.1 | XP_008763900.1 | 4 - 5 | 4833 | 78 | ||
GenBank mRNA | S76466.1 | - | 4 - 5 | 828 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |