Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201655.1 | NP_970617.1 | 1 - 2 | 184 | 74 | |
XM_008772274.1 | XP_008770496.1 | 1 - 2 | 461 | 74 | ||
XM_008772275.1 | XP_008770497.1 | 1 - 2 | 461 | 74 | ||
XM_008772276.1 | XP_008770498.1 | 1 - 2 | 461 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |