Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017286.2 | NP_058982.1 | 6 - 7 | 1147 | 125 | |
GenBank mRNA | BC089100.1 | - | 6 - 7 | 1172 | 7307104 | 125 |
J05156.1 | - | 6 - 7 | 1167 | 125 | ||
M22615.1 | - | 1 - 2 | 118 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |