Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019161.2 | NP_062034.2 | 2 - 3 | 649 | 78 | |
GenBank mRNA | D83348.1 | - | 2 - 3 | 655 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |