Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022289.3 | NP_071625.2 | 2 - 3 | 664 | 102 | |
XM_006232135.2 | XP_006232197.1 | 2 - 3 | 677 | 102 | ||
GenBank mRNA | AF068746.2 | - | 1 - 2 | 376 | 102 | |
AF305780.1 | - | 2 - 3 | 650 | 102 | ||
BC061554.1 | - | 2 - 3 | 668 | 6890629 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |