Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031020.2 | NP_112282.2 | 3 - 4 | 606 | 133 | |
XM_006256142.2 | XP_006256204.1 | 3 - 4 | 318 | 133 | ||
GenBank mRNA | AB000783.1 | - | 3 - 4 | 605 | 133 | |
AF346293.1 | - | 3 - 4 | 303 | 133 | ||
BC092193.1 | - | 3 - 4 | 606 | 7313217 | 133 | |
U73142.1 | - | 3 - 4 | 133 | |||
U91847.1 | - | 3 - 4 | 303 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |