Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 1 - 2 | 641 | 68 | |
XM_006241018.2 | XP_006241080.1 | 2 - 3 | 802 | 68 | ||
XM_006241019.2 | XP_006241081.1 | 2 - 3 | 775 | 68 | ||
XM_006241020.2 | XP_006241082.1 | 2 - 3 | 800 | 68 | ||
GenBank mRNA | AF029109.1 | - | 1 - 2 | 641 | 68 | |
BC161842.1 | - | 2 - 3 | 703 | 9102177 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |