Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053988.1 | NP_446440.1 | 2 - 3 | 224 | 148 | |
XM_008772466.1 | XP_008770688.1 | 3 - 4 | 403 | 148 | ||
GenBank mRNA | BC087603.1 | - | 2 - 3 | 194 | 7312595 | 148 |
X66974.1 | - | 2 - 3 | 224 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |